Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029868.2 | NP_001025039.1 | 2 | 859 | ||
GenBank mRNA | AK040378.1 | - | 1 | 1289 | ||
AK084004.1 | - | 1 | 1361 | |||
AK165004.1 | - | 1 | 1441 | |||
BC056462.1 | - | 2 | 205 | 5703229 |