Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030307.2 | NP_001025478.1 | 10 | 1203 | ||
GenBank mRNA | AK136418.1 | - | 10 | 988 | ||
AK144139.1 | - | 10 | 991 | |||
AK166105.1 | - | 10 | 979 | |||
AK167973.1 | - | 10 | 954 | |||
BC068171.1 | - | 10 | 959 | |||
BC099966.1 | - | 10 | 1043 | 6435181 |