Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005247.2 | NP_001005247.2 | 19 | 3223 | ||
NM_001005248.2 | NP_001005248.2 | 19 | 3199 | |||
NM_001167864.1 | NP_001161336.1 | 19 | 3124 | |||
GenBank mRNA | AF534396.1 | - | 19 | 3020 | ||
AF534397.1 | - | 19 | 2996 | |||
AF534398.1 | - | 17 | 2585 | |||
AF534399.1 | - | 20 | 3151 | |||
AK129265.1 | - | 19 | 3146 | |||
AK140780.1 | - | 8 | 1405 | |||
AK141088.1 | - | 18 | 3848 | |||
AK154612.1 | - | 19 | 2952 | |||
AK157182.1 | - | 19 | 2990 | |||
AK161039.1 | - | 10 | 1594 | |||
BC046405.1 | - | 14 | 2202 | |||
BC046443.1 | - | 19 | 2837 | |||
BC082542.1 | - | 19 | 3124 | 6405811 |