Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133902.2 | NP_598663.2 | 8 | 1008 | ||
GenBank mRNA | AF328927.1 | - | 9 | 1275 | ||
AK014487.1 | - | 5 | 624 | |||
BC022601.1 | - | 8 | 1008 | 4160199 |