Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146555.1 | NP_666766.1 | 1 | 282 | ||
GenBank mRNA | BC111816.1 | - | 1 | 281 |