Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146649.2 | NP_666860.2 | 1 | 272 |
Gene Symbol: Olfr1154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146647.2 | NP_666858.2 | 1 | 245 |