Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146671.1 | NP_666882.1 | 1 | 658 | ||
GenBank mRNA | BC127972.1 | - | 1 | 662 | 40135122 |