Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146939.1 | NP_667150.1 | 1 | 511 | ||
GenBank mRNA | BC119279.1 | - | 1 | 548 | 8734028 | |
BC119283.1 | - | 1 | 548 | 8734032 |