Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146951.1 | NP_667162.1 | 1 | 439 | ||
GenBank mRNA | BC109141.1 | - | 1 | 439 | 40041785 | |
BC109142.1 | - | 1 | 438 |