Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147024.2 | NP_667235.1 | 2 | 800 | ||
GenBank mRNA | BC139813.1 | - | 1 | 669 | 40135620 |