Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 3 | 1064 | ||
GenBank mRNA | AK045508.1 | - | 3 | 1065 | ||
AK051262.1 | - | 3 | 1065 | |||
AK122276.1 | - | 3 | 712 |