Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080448.4 | NP_536696.4 | 6 | 1558 | ||
GenBank mRNA | AF338472.1 | - | 5 | 716 | ||
AF496547.1 | - | 6 | 788 | |||
AK045508.1 | - | 6 | 1559 | |||
AK051262.1 | - | 6 | 1559 | |||
AK122276.1 | - | 6 | 1206 |