Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fev | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153111.2 | NP_694751.1 | 1, 2 | 256 | ||
GenBank mRNA | AY049085.1 | - | 1, 2 | 266 | ||
BC138435.1 | - | 1, 2 | 171 | 8861455 | ||
BC138436.1 | - | 1, 2 | 171 | 8861456 |