Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fev | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153111.2 | NP_694751.1 | 3 | 543 | ||
GenBank mRNA | AY049085.1 | - | 3 | 553 | ||
BC138435.1 | - | 3 | 458 | 8861455 | ||
BC138436.1 | - | 3 | 458 | 8861456 | ||
BC145276.1 | - | 3 | 490 |