Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252067.1 | NP_001238996.1 | 16 | 1927 | ||
NM_173048.3 | NP_766636.2 | 17 | 2161 | |||
GenBank mRNA | AK031086.1 | - | 17 | 2133 | ||
AK082327.1 | - | 16 | 1891 | |||
AK122212.2 | - | 17 | 2130 | |||
AK132183.1 | - | 17 | 2129 | |||
AK147882.1 | - | 17 | 2129 | |||
AK193796.1 | - | 1 | 57 | |||
BC024266.1 | - | 2768 | ||||
BC025051.1 | - | 4 | 562 | |||
BC056424.1 | - | 17 | 2050 | |||
BC085617.1 | - | 12 | 4640 | |||
BC110676.1 | - | 14 | 2459 |