Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB221554.1 | - | 2 | 435 | ||
AF149820.1 | - | 2 | 429 | |||
AF177755.1 | - | 2 | 417 | |||
AF177756.1 | - | 2 | 402 | |||
AF177757.1 | - | 2 | 1275 | |||
AK035490.1 | - | 2 | 432 | |||
AK076482.1 | - | 2 | 325 | |||
AK141466.1 | - | 2 | 325 | |||
AK145553.1 | - | 2 | 327 | |||
AK157424.1 | - | 2 | 424 | |||
BC016573.1 | - | 2 | 817 | |||
BC147770.1 | - | 2 | 527 | 9087493 | ||
BC147784.1 | - | 2 | 527 | 9087503 | ||
CK624100.1 | - | 2 | 313 | |||
DV655694.1 | - | 2 | 395 |