Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA407185.1 | - | 1 | 107 | ||
AB221554.1 | - | 1 | 90 | |||
AF149820.1 | - | 1 | 84 | |||
AF177757.1 | - | 1 | 930 | |||
BC016573.1 | - | 1 | 472 | |||
BC147770.1 | - | 1 | 182 | 9087493 | ||
BC147784.1 | - | 1 | 182 | 9087503 | ||
CA320790.1 | - | 1 | 517 |