Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011932.2 | NP_036062.1 | 1 | 153 | ||
GenBank mRNA | AF163255.1 | - | 1 | 107 | ||
AF186023.1 | - | 1 | 128 | |||
AK079794.1 | - | 1 | 142 | |||
AK145745.1 | - | 1 | 119 | |||
AK146180.1 | - | 1 | 122 | |||
AK169584.1 | - | 1 | 141 | |||
BC014759.1 | - | 1 | 149 | 4486834 |