Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256042.1 | NP_001242971.1 | 4 | 496 | ||
NM_001256044.1 | NP_001242973.1 | 3 | 387 | |||
NM_011939.3 | NP_036069.1 | 4 | 496 | |||
GenBank mRNA | AB029349.1 | - | 4 | 428 | ||
AB029350.1 | - | 4 | 423 | |||
AF160966.1 | - | 4 | 423 | |||
AK148056.1 | - | 3 | 359 | |||
AK189360.1 | - | 1 | 69 | |||
AK216507.1 | - | 1 | 69 | |||
BC138130.1 | - | 4 | 496 | 8861149 | ||
BC145153.1 | - | 4 | 496 | |||
BC145154.1 | - | 4 | 496 |