Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170756.2 | NP_739562.1 | 2 | 376 | ||
GenBank mRNA | AK051741.1 | - | 2 | 322 | ||
AK122364.1 | - | 1 | 440 | |||
AK144826.1 | - | 2 | 348 | |||
AK156188.1 | - | 2 | 377 | |||
AK162743.1 | - | 2 | 376 | |||
AK170356.1 | - | 2 | 377 | |||
AK171240.1 | - | 2 | 350 | |||
AK171664.1 | - | 2 | 354 | |||
AK171852.1 | - | 2 | 366 | |||
AK201572.1 | - | 1 | 105 | |||
AK210472.1 | - | 1 | 104 | |||
BC034597.1 | - | 2 | 365 | 5363513 |