Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153599.3 | NP_705827.2 | 4 | 369 | ||
GenBank mRNA | AK131948.1 | - | 4 | 308 | ||
AK182034.1 | - | 2 | 121 | |||
BI151839.1 | - | 4 | 369 | |||
CF723869.1 | - | 2 | 71 | |||
CF730982.1 | - | 4 | 287 |