Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168508.1 | NP_001161980.1 | 8 | 1062 | ||
NM_001168513.1 | NP_001161985.1 | 8 | 663 | |||
NM_001168514.1 | NP_001161986.1 | 9 | 1107 | |||
NM_011951.3 | NP_036081.1 | 8 | 1062 | |||
GenBank mRNA | AF128892.1 | - | 8 | 960 | ||
AK089059.1 | - | 8 | 976 | |||
AK133684.1 | - | 8 | 975 | |||
AK151348.1 | - | 8 | 664 | |||
AK153025.1 | - | 8 | 664 | |||
BC012235.1 | - | 8 | 1062 | 4195842 | ||
BC018227.1 | - | 9 | 1004 | |||
BC085303.1 | - | 8 | 1038 | 5389457 | ||
CX568915.1 | - | 7 | 456 | |||
D83073.1 | - | 8 | 893 | |||
U10871.1 | - | 8 | 960 |