Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030595.1 | NP_085098.1 | 53, 54 | 8566 | ||
GenBank mRNA | AF072372.1 | - | 11, 12 | 1445 | ||
AK043125.1 | - | 14, 15 | 1842 | |||
BC034103.1 | - | 3, 4 | 410 | |||
BC064452.1 | - | 3, 4 | 377 | |||
Y18276.1 | - | 53, 54 | 8566 |