Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007676.4 | NP_031702.3 | 2 | 593 | ||
GenBank mRNA | AB500066.1 | - | 1 | 169 | ||
BC030357.1 | - | 2 | 603 | 5367680 | ||
U34272.1 | - | 2 | 549 |