Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013711.3 | NP_038739.2 | 2, 3 | 211 | ||
GenBank mRNA | AB027566.1 | - | 2, 3 | 211 | ||
AF136399.1 | - | 2, 3 | 214 | |||
AF171053.1 | - | 2, 3 | 177 | |||
AF412308.1 | - | 2, 3 | 213 | |||
AK008004.1 | - | 2, 3 | 130 | |||
AK075828.1 | - | 2, 3 | 189 | |||
BC013688.1 | - | 2, 3 | 191 | 3584154 | ||
BC029344.1 | - | 2, 3 | 191 | |||
BC052157.1 | - | 2, 3 | 190 | 6506286 | ||
BC052758.1 | - | 2, 3 | 190 |