Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acmsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033041.2 | NP_001028213.1 | 3 | 211 | ||
GenBank mRNA | AB071419.1 | - | 3 | 138 | ||
AK143962.1 | - | 3 | 213 | |||
BC139281.1 | - | 3 | 162 | 8862303 |