Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024195.2 | NP_077157.2 | 2, 3 | 375 | ||
GenBank mRNA | AF338818.1 | - | 1, 2 | 123 | ||
AK012855.1 | - | 1, 2 | 15 | |||
AK034878.1 | - | 2, 3 | 319 | |||
AK090159.1 | - | 2, 3 | 377 | |||
AK135261.1 | - | 1, 2 | 26 | |||
AK157312.1 | - | 2, 3 | 355 | |||
AK167436.1 | - | 1, 2 | 73 | |||
AK169937.1 | - | 2, 3 | 349 | |||
AK169995.1 | - | 2, 3 | 347 | |||
AK172252.1 | - | 2, 3 | 304 | |||
AY321368.1 | - | 1, 2 | 123 | |||
AY321369.1 | - | 1, 2 | 123 | |||
BC025438.1 | - | 2, 3 | 362 | 5038563 |