Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sowahc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172939.3 | NP_766527.3 | 1 | 632 | ||
GenBank mRNA | AK030944.1 | - | 1 | 633 |
Gene Symbol: Sept10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028519.1 | - | 1126 |