Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172945.2 | NP_766533.2 | 8 | 1002 | ||
GenBank mRNA | BC056928.1 | - | 3 | 231 | ||
BC071189.1 | - | 2 | 88 | |||
BC098207.1 | - | 1 | 41 | |||
BC151158.1 | - | 8 | 980 | 9088046 | ||
BC172028.1 | - | 8 | 981 | |||
CA315262.1 | - | 7 | 688 | |||
CD578820.1 | - | 2 | 100 |