Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021477.5 | NP_067452.2 | 7, 8 | 1735 | ||
NM_183188.2 | NP_899011.2 | 3, 4 | 630 | |||
GenBank mRNA | AB041596.2 | - | 6, 7 | 1042 | ||
AF107204.1 | - | 3, 4 | 418 | |||
AF191501.1 | - | 3, 4 | 631 | |||
AK134833.1 | - | 5, 6 | 666 | |||
AY659954.1 | - | 3, 4 | 406 | |||
AY659955.1 | - | 3, 4 | 406 | |||
AY659956.1 | - | 3, 4 | 466 | |||
AY659957.1 | - | 3, 4 | 466 | |||
AY659958.1 | - | 3, 4 | 466 | |||
BC053009.1 | - | 7, 8 | 1735 | |||
BC059002.1 | - | 6, 7 | 1399 | 6821627 | ||
EF410136.1 | - | 5, 6 | 786 |