Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013385.2 | NP_001013403.1 | 9 | 2850 | ||
GenBank mRNA | BC080284.1 | - | 9 | 2621 | 6402528 |
Set Membership: |
G-protein coupled receptor |