Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081315.1 | NP_001074784.1 | 11 | 3452 | ||
GenBank mRNA | AK129322.1 | - | 9 | 1707 | ||
AK147555.1 | - | 11 | 3451 | |||
AK162638.1 | - | 4 | 300 | |||
BC157915.1 | - | 10 | 3217 | 9088158 |