Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033367.3 | NP_001028539.1 | 4 | 530 | ||
GenBank mRNA | AK143864.1 | - | 4 | 531 | ||
AK171624.1 | - | 4 | 525 | |||
BC144861.1 | - | 2 | 301 | |||
BC144862.1 | - | 2 | 287 | |||
BC145617.1 | - | 3 | 324 |