Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ss18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161369.1 | NP_001154841.1 | 5 | 661 | ||
NM_009280.2 | NP_033306.2 | 5 | 661 | |||
GenBank mRNA | AK039430.1 | - | 5 | 511 | ||
AK083622.1 | - | 5 | 523 | |||
AK083887.1 | - | 5 | 545 | |||
AK149325.1 | - | 5 | 523 | |||
AK166053.1 | - | 5 | 514 | |||
AK178758.1 | - | 2 | 185 | |||
BC065062.1 | - | 5 | 498 | |||
BC096742.1 | - | 5 | 498 | 6416415 | ||
X93357.1 | - | 5 | 661 |