Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012009.4 | NP_036139.3 | 397 | |||
GenBank mRNA | AB196832.1 | - | 345 | |||
AB196833.1 | - | 345 | ||||
AB196834.1 | - | 345 | ||||
AB196835.1 | - | 345 | ||||
AF020263.1 | - | 345 | ||||
AK089191.1 | - | 402 | ||||
BC116698.1 | - | 397 | 40105516 | |||
BC117508.1 | - | 397 | 40105515 | |||
BC145540.1 | - | 403 | 9054103 | |||
BC171965.1 | - | 403 | 9054251 | |||
DQ131180.1 | - | 345 | ||||
DQ132507.1 | - | 345 | ||||
DQ132508.1 | - | 345 | ||||
DQ132509.1 | - | 345 | ||||
DQ132510.1 | - | 345 |