Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160420.2 | NP_001153892.1 | 4 | 514 | ||
NM_001160421.2 | NP_001153893.1 | 3 | 625 | |||
NM_001160422.1 | NP_001153894.1 | 3 | 625 | |||
NM_054076.3 | NP_473417.2 | 3 | 625 | |||
GenBank mRNA | AF333980.1 | - | 3 | 624 | ||
AK053318.1 | - | 3 | 408 | |||
AK153750.1 | - | 4 | 515 | |||
AY077682.1 | - | 3 | 617 | |||
BC099435.1 | - | 3 | 458 | 30788813 | ||
BC115928.1 | - | 3 | 578 | 40104738 |