Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf512b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164597.1 | NP_001158069.1 | 5 | 944 | ||
GenBank mRNA | AK129308.1 | - | 5 | 944 | ||
BC056460.1 | - | 3 | 931 |