Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf512b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164597.1 | NP_001158069.1 | 14 | 2225 | ||
GenBank mRNA | AK129308.1 | - | 14 | 2225 | ||
BC056460.1 | - | 12 | 2212 |