Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175651.4 | NP_783582.1 | 2 | 262 | ||
GenBank mRNA | AK035828.1 | - | 2 | 263 | ||
AK035938.1 | - | 1 | 802 | |||
AK139155.1 | - | 1 | 948 | |||
AK162570.1 | - | 1 | 751 | |||
BC096464.1 | - | 187 | 6390940 |