Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175651.4 | NP_783582.1 | 2, 3 | 352 | ||
GenBank mRNA | AK035828.1 | - | 2, 3 | 353 | ||
AK035938.1 | - | 1, 2 | 892 | |||
AK139155.1 | - | 1, 2 | 1038 | |||
AK162570.1 | - | 1, 2 | 841 | |||
BC096464.1 | - | 277 | 6390940 |