Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290528.1 | NP_001277457.1 | 8, 9 | 2370 | ||
GenBank mRNA | BC046514.1 | - | 5, 6 | 766 | ||
BC057197.1 | - | 7, 8 | 1079 | |||
BC078445.1 | - | 8, 9 | 1409 |