Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178752.3 | NP_848867.2 | 2 | 1122 | ||
GenBank mRNA | AK084531.1 | - | 2 | 1108 | ||
AK086166.1 | - | 2 | 1125 | |||
AK143704.1 | - | 2 | 1148 | |||
AK166345.1 | - | 2 | 1153 | |||
BC037479.1 | - | 2 | 1099 | 4458984 |