Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178752.3 | NP_848867.2 | 2 | 930 | ||
GenBank mRNA | AK084531.1 | - | 2 | 916 | ||
AK085577.1 | - | 937 | ||||
AK086166.1 | - | 2 | 933 | |||
AK143704.1 | - | 2 | 956 | |||
AK166345.1 | - | 2 | 961 | |||
BC037479.1 | - | 2 | 907 | 4458984 |