Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173012.3 | NP_766600.2 | 11 | 1825 | ||
GenBank mRNA | AW048776.1 | - | 1 | 287 | ||
BC050979.1 | - | 7 | 730 | |||
BC055685.1 | - | 11 | 1825 | 6403034 |