Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177461.1 | NP_803412.1 | 12 | 2541 | ||
GenBank mRNA | AK076763.1 | - | 6 | 1198 | ||
AK077217.1 | - | 5 | 1369 | |||
AK129419.1 | - | 12 | 2279 | |||
AK147587.1 | - | 12 | 2342 | |||
AK147646.1 | - | 12 | 2554 |