Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177771.5 | NP_808439.2 | 9 | 1417 | ||
GenBank mRNA | AK173029.1 | - | 9 | 1417 | ||
BC025563.1 | - | 9 | 1428 | 5322893 | ||
BC065167.1 | - | 8 | 1113 | |||
CF739377.1 | - | 3 | 285 |