Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177771.5 | NP_808439.2 | 1 | 214 | ||
GenBank mRNA | AK173029.1 | - | 1 | 214 | ||
BC025563.1 | - | 1 | 210 | 5322893 | ||
BC065167.1 | - | 1 | 110 | |||
BY195998.1 | - | 1 | 135 | |||
BY248410.1 | - | 1 | 134 |