Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128080.2 | NP_001121552.1 | 4 | 393 | ||
NM_001128081.1 | NP_001121553.1 | 3 | 508 | |||
NM_001128082.1 | NP_001121554.1 | 3 | 508 | |||
NM_020286.3 | NP_064682.1 | 3 | 508 | |||
GenBank mRNA | AF175771.1 | - | 3 | 357 | ||
AF291425.1 | - | 3 | 476 | |||
AJ279791.1 | - | 3 | 468 | |||
AJ279792.1 | - | 3 | 468 | |||
AJ279793.1 | - | 3 | 468 | |||
AJ279794.1 | - | 3 | 468 | |||
AJ279795.1 | - | 3 | 468 | |||
AK013248.1 | - | 3 | 284 | |||
AK154694.1 | - | 3 | 529 | |||
AK171199.1 | - | 3 | 508 | |||
AY039000.1 | - | 4 | 401 | |||
BC051204.1 | - | 3 | 498 | |||
BC055858.1 | - | 3 | 482 | 4913671 |