Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013848.2 | NP_038876.2 | 5 | 904 | ||
GenBank mRNA | AF153906.1 | - | 5 | 869 | ||
AK036702.1 | - | 5 | 869 | |||
AK158114.1 | - | 5 | 887 | |||
BC063257.1 | - | 5 | 838 | 6410069 |