Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012056.2 | NP_036186.2 | 5 | 886 | ||
GenBank mRNA | AF090334.1 | - | 5 | 812 | ||
AF279263.1 | - | 5 | 864 | |||
AK007499.1 | - | 3 | 276 | |||
AK189046.1 | - | 2 | 119 | |||
BC026133.1 | - | 5 | 826 | 4018235 | ||
BC043129.1 | - | 5 | 886 | 6810029 |